Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hmx2
Entrez Gene ID
15372
Gene Name
H6 homeobox 2
Gene Aliases
Nkx-5.2, Nkx5-2
Chromosome Location
Chr.7: 131548773 - 131558867 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC023402.1 - 1 - 2 116 85

View Full Product Details