Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Avil
Entrez Gene ID
11567
Gene Name
advillin
Gene Aliases
Advil, DOC6
Chromosome Location
Chr.10: 127000075 - 127020994 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009635.3 NP_033765.2 11 - 12 1337 81
GenBank mRNA AF041448.1 - 11 - 12 1288 81
AF059486.1 - 11 - 12 1307 81
AK154851.1 - 11 - 12 1308 81
AK155900.1 - 11 - 12 1308 81
AK170878.1 - 11 - 12 1308 81
AK171319.1 - 11 - 12 1309 81
AK171324.1 - 11 - 12 1308 81
AK204210.1 - 2 - 3 218 81
BC119222.1 - 11 - 12 1242 81
BC120545.1 - 11 - 12 1242 81

View Full Product Details