The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016762.2 | NP_058042.2 | 15 - 16 | 2761 | 77 |
GenBank mRNA | AK138211.1 | - | 15 - 16 | 2758 | 77 |
AK163818.1 | - | 15 - 16 | 2720 | 77 | |
BC005429.1 | - | 15 - 16 | 2631 | 77 | |
BC092298.1 | - | 15 - 16 | 2704 | 77 | |
U69262.1 | - | 15 - 16 | 2760 | 77 |