Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Orc5
Entrez Gene ID
26429
Gene Name
origin recognition complex, subunit 5
Gene Aliases
AL033327, MmORC5, Orc5l
Chromosome Location
Chr.5: 22486484 - 22550367 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011959.2 NP_036089.1 13 - 14 1330 115
GenBank mRNA AJ007360.1 - 13 - 14 1327 115
AK084683.1 - 13 - 14 1358 115
AK144983.1 - 13 - 14 1346 115
AK165912.1 - 13 - 14 1336 115
AK166537.1 - 13 - 14 1269 115
AK169683.1 - 12 - 13 1385 115
BC006927.1 - 13 - 14 1346 115

View Full Product Details