Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyrm1
Entrez Gene ID
73919
Gene Name
LYR motif containing 1
Gene Aliases
1110065L10Rik, 2310004B22Rik, 4930404J24Rik
Chromosome Location
Chr.7: 119895807 - 119956203 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK015083.1 - 2 - 3 226 77
BC011339.1 - 1 - 2 208 77

View Full Product Details