Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trappc10
Entrez Gene ID
216131
Gene Name
trafficking protein particle complex 10
Gene Aliases
B230307C21Rik, Gm870, Tmem1
Chromosome Location
Chr.10: 78186725 - 78244642 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081055.1 NP_001074524.1 1 - 2 207 91
GenBank mRNA AK154290.1 - 1 - 2 143 91
AK166500.1 - 1 - 2 207 91

View Full Product Details