Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpn2
Entrez Gene ID
71756
Gene Name
carboxypeptidase N, polypeptide 2
Gene Aliases
1300018K11Rik
Chromosome Location
Chr.16: 30256378 - 30267614 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027904.3 NP_082180.2 1 - 2 79 83
GenBank mRNA AK005049.1 - 1 - 2 81 83
BC138287.1 - 1 - 2 166 83
BC138288.1 - 1 - 2 166 83

View Full Product Details