Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nsun3
Entrez Gene ID
106338
Gene Name
NOL1/NOP2/Sun domain family member 3
Gene Aliases
6720484A09Rik, AU022521
Chromosome Location
Chr.16: 62732444 - 62786794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK032127.1 - 1 - 2 88 81
AK153942.1 - 1 - 2 108 81
AK169841.1 - 1 - 2 97 81

View Full Product Details