The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175535.3 | NP_780744.2 | 13 - 14 | 1752 | 94 |
GenBank mRNA | AK129345.1 | - | 10 - 11 | 1241 | 94 |