Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgap20
Entrez Gene ID
244867
Gene Name
Rho GTPase activating protein 20
Gene Aliases
6530403F17Rik, A530023E23Rik, RARhoGAP, mKIAA1391
Chromosome Location
Chr.9: 51765087 - 51853856 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175535.3 NP_780744.2 5 - 6 696 95
GenBank mRNA AK040756.1 - 5 - 6 696 95
AK129345.1 - 2 - 3 185 95

View Full Product Details