Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc35f2
Entrez Gene ID
72022
Gene Name
solute carrier family 35, member F2
Gene Aliases
1500009K05Rik, AU019213
Chromosome Location
Chr.9: 53771226 - 53818161 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028060.3 NP_082336.3 5 - 6 978 82
GenBank mRNA AK005182.1 - 5 - 6 774 82
BC019202.1 - 5 - 6 779 82
BC055843.1 - 5 - 6 975 82

View Full Product Details