Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nccrp1
Entrez Gene ID
233038
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
1190020J12Rik, Gm163, NCCRP-1
Chromosome Location
Chr.7: 28543596 - 28547254 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081115.1 NP_001074584.1 5 - 6 855 107
GenBank mRNA AK028022.1 - 5 - 6 505 107

View Full Product Details