The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177475.3 | NP_803426.2 | 1 - 2 | 142 | 53 |
GenBank mRNA | AK031331.1 | - | 1 - 2 | 40 | 53 |
AK144644.1 | - | 1 - 2 | 100 | 53 | |
AK144736.1 | - | 1 - 2 | 100 | 53 | |
BC094574.1 | - | 1 - 2 | 71 | 53 |