The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177794.3 | NP_808462.2 | 2 - 3 | 425 | 69 |
GenBank mRNA | AK028283.1 | - | 2 - 3 | 426 | 69 |
AK084351.1 | - | 2 - 3 | 403 | 69 | |
AK143866.1 | - | 2 - 3 | 403 | 69 | |
BC117503.1 | - | 2 - 3 | 347 | 69 |