The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177794.3 | NP_808462.2 | 5 - 6 | 835 | 81 |
GenBank mRNA | AK028283.1 | - | 5 - 6 | 836 | 81 |
AK084351.1 | - | 5 - 6 | 813 | 81 | |
AK143866.1 | - | 5 - 6 | 813 | 81 | |
BC117502.1 | - | 6 - 7 | 839 | 81 | |
BC117503.1 | - | 5 - 6 | 757 | 81 |