Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Eaf2
- Entrez Gene ID
-
106389
- Gene Name
- ELL associated factor 2
- Gene Aliases
-
AW048865, FESTA-L, FESTA-S, Festa, Traits, U19
- Chromosome Location
-
Chr.16: 36792884 - 36874826 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK076871.1 |
- |
5 - 6 |
668 |
125 |
View Full Product Details