The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033498.1 | NP_001028670.1 | 1 - 2 | 332 | 59 |
GenBank mRNA | AK040545.1 | - | 2 - 3 | 221 | 59 |
AK041631.1 | - | 1 - 2 | 332 | 59 | |
BC138948.1 | - | 1 - 2 | 149 | 59 | |
BC138949.1 | - | 1 - 2 | 149 | 59 |