Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gramd2
Entrez Gene ID
546134
Gene Name
GRAM domain containing 2
Gene Aliases
BC064463
Chromosome Location
Chr.9: 59679462 - 59718871 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033498.1 NP_001028670.1 7 - 8 805 107
GenBank mRNA AK040545.1 - 8 - 9 694 107
AK041631.1 - 7 - 8 805 107
BC138948.1 - 7 - 8 622 107
BC138949.1 - 7 - 8 622 107

View Full Product Details