Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rmi2
- Entrez Gene ID
-
223970
- Gene Name
- RMI2, RecQ mediated genome instability 2, homolog (S. cerevisiae)
- Gene Aliases
-
A630055G03Rik, Gm537, Gm929
- Chromosome Location
-
Chr.16: 10835059 - 10843235 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK080331.1 |
- |
- |
326 |
81 |
View Full Product Details