The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029714.3 | NP_083990.2 | 15 - 16 | 1859 | 85 |
GenBank mRNA | AK077059.1 | - | 15 - 16 | 1841 | 85 |
AK138347.1 | - | 11 - 12 | 1365 | 85 | |
BC144969.1 | - | 15 - 16 | 1839 | 85 | |
GQ225581.1 | - | 14 - 15 | 1760 | 85 |