Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fcgbp
Entrez Gene ID
215384
Gene Name
Fc fragment of IgG binding protein
Gene Aliases
A430096B05Rik
Chromosome Location
Chr.7: 28071236 - 28120864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122603.1 NP_001116075.1 19 - 20 7760 60
GenBank mRNA AK040441.1 - 9 - 10 4464 60
BC024494.1 - 2 - 3 454 60
BC024520.1 - 4 - 5 1422 60
BC024554.1 - 4 - 5 925 60
BC026653.1 - 5 - 6 1965 60
BC029250.1 - 7 - 8 2782 60
BC030871.1 - 8 - 9 3609 60
BC036158.1 - 5 - 6 1802 60
BC036159.1 - 4 - 5 1084 60
BC039286.1 - 10 - 11 4186 60
BC048952.1 - 3 - 4 956 60
BC055475.1 - 5 - 6 1473 60
BC069956.1 - 5 - 6 1768 60
BC092067.1 - 5 - 6 1839 60
BC100564.1 - 2 - 3 353 60
EH108930.1 - 1 - 2 60

View Full Product Details