Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcf3
Entrez Gene ID
21423
Gene Name
transcription factor 3
Gene Aliases
A1, AA408400, ALF2, AW209082, E12, E12/E47, E2A, E47, ME2, Pan1, Pan2, TCF-3, Tcfe2a, VDIR, bHLHb21
Chromosome Location
Chr.10: 80409165 - 80433653 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164147.1 NP_001157619.1 10 - 11 1202 77
NM_001164148.1 NP_001157620.1 10 - 11 1202 77
NM_001164149.1 NP_001157621.1 10 - 11 1199 77
NM_001164150.1 NP_001157622.1 10 - 11 1202 77
NM_001164151.1 NP_001157623.1 10 - 11 1199 77
NM_001164152.1 NP_001157624.1 10 - 11 1202 77
NM_001164153.1 NP_001157625.1 10 - 11 1199 77
NM_011548.4 NP_035678.3 10 - 11 1199 77
GenBank mRNA AF352579.1 - 10 - 11 894 77
AK017617.1 - 10 - 11 1167 77
AK034777.1 - 10 - 11 1182 77
AK038738.1 - 10 - 11 1186 77
AK146786.1 - 10 - 11 1193 77
AK147735.1 - 9 - 10 858 77
AK156265.1 - 10 - 11 1182 77
AK161009.1 - 10 - 11 1188 77
AK170674.1 - 10 - 11 1197 77
BC006860.1 - 10 - 11 1153 77
BC018260.1 - 9 - 10 827 77

View Full Product Details