Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcf3
Entrez Gene ID
21423
Gene Name
transcription factor 3
Gene Aliases
A1, AA408400, ALF2, AW209082, E12, E12/E47, E2A, E47, ME2, Pan1, Pan2, TCF-3, Tcfe2a, VDIR, bHLHb21
Chromosome Location
Chr.10: 80409165 - 80433653 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164147.1 NP_001157619.1 13 - 14 1484 101
NM_001164148.1 NP_001157620.1 13 - 14 1484 101
NM_001164149.1 NP_001157621.1 13 - 14 1481 101
NM_001164150.1 NP_001157622.1 13 - 14 1484 101
NM_001164151.1 NP_001157623.1 13 - 14 1481 101
NM_001164152.1 NP_001157624.1 13 - 14 1484 101
NM_001164153.1 NP_001157625.1 13 - 14 1481 101
NM_011548.4 NP_035678.3 13 - 14 1481 101
GenBank mRNA AF352579.1 - 13 - 14 1176 101
AK017617.1 - 13 - 14 1449 101
AK034777.1 - 13 - 14 1464 101
AK038738.1 - 13 - 14 1468 101
AK146786.1 - 13 - 14 1476 101
AK147735.1 - 12 - 13 1140 101
AK156265.1 - 13 - 14 1464 101
AK161009.1 - 13 - 14 1470 101
AK170674.1 - 13 - 14 1479 101
BC006860.1 - 13 - 14 1435 101
BC018260.1 - 12 - 13 1109 101

View Full Product Details