Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcf3
Entrez Gene ID
21423
Gene Name
transcription factor 3
Gene Aliases
A1, AA408400, ALF2, AW209082, E12, E12/E47, E2A, E47, ME2, Pan1, Pan2, TCF-3, Tcfe2a, VDIR, bHLHb21
Chromosome Location
Chr.10: 80409165 - 80433653 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164150.1 NP_001157622.1 18 - 19 2204 84
NM_001164151.1 NP_001157623.1 18 - 19 2201 84
NM_001164152.1 NP_001157624.1 18 - 19 2201 84
NM_001164153.1 NP_001157625.1 18 - 19 2198 84
NM_011548.4 NP_035678.3 18 - 19 2198 84
GenBank mRNA AF352579.1 - 18 - 19 1893 84
AK034777.1 - 18 - 19 2180 84
AK038738.1 - 18 - 19 2188 84
AK147735.1 - 17 - 18 1861 84
AK161009.1 - 18 - 19 2190 84
BC006860.1 - 18 - 19 2152 84
D16631.1 - 4 - 5 653 84
X17500.1 - 5 - 6 706 84

View Full Product Details