Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp30
Entrez Gene ID
22693
Gene Name
zinc finger protein 30
Gene Aliases
2610306P15Rik, Zfp-30
Chromosome Location
Chr.7: 29784790 - 29794540 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013705.1 NP_038733.1 1 - 2 118 94
GenBank mRNA AK032058.1 - - 826 94
AK173073.1 - - 864 94
AY605122.1 - 1 - 2 118 94
BC049120.1 - 1 - 2 119 94
BC051972.1 - 1 - 2 318 94
Z30174.1 - 1 - 2 118 94

View Full Product Details