The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009795.3 | NP_033925.2 | 5 - 6 | 449 | 81 |
GenBank mRNA | AK028092.1 | - | 3 - 4 | 195 | 81 |
AK132819.1 | - | 4 - 5 | 259 | 81 | |
BC018352.1 | - | 5 - 6 | 443 | 81 | |
BC090988.1 | - | 5 - 6 | 449 | 81 |