The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172898.3 | NP_766486.1 | 1 - 2 | 720 | 68 |
GenBank mRNA | AK049284.1 | - | 1 - 2 | 721 | 68 |
AK083069.1 | - | 1 - 2 | 861 | 68 | |
BC052773.1 | - | 1 - 2 | 183 | 68 | |
BC158021.1 | - | 1 - 2 | 228 | 68 |