Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clmp
Entrez Gene ID
71566
Gene Name
CXADR-like membrane protein
Gene Aliases
9030425E11Rik, ACAM, ASP5, AW557819
Chromosome Location
Chr.9: 40685964 - 40785553 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133733.4 NP_598494.2 6 - 7 103
GenBank mRNA AB040490.1 - 6 - 7 1107 103
AK018527.1 - 1 - 2 103
AK028983.1 - 6 - 7 103
AK033723.1 - 6 - 7 103
AY259213.1 - 7 - 8 819 103
AY326421.1 - 6 - 7 825 103
BC026447.1 - 6 - 7 103

View Full Product Details