Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Elmod1
Entrez Gene ID
270162
Gene Name
ELMO/CED-12 domain containing 1
Gene Aliases
4831417L10, AI428506, AI834991
Chromosome Location
Chr.9: 53911457 - 53975310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177769.4 NP_808437.2 10 - 11 1162 66
GenBank mRNA AK132239.1 - 10 - 11 1163 66
AK164094.1 - - 314 66
BC049160.1 - 8 - 9 822 66

View Full Product Details