Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpusd4
Entrez Gene ID
71989
Gene Name
RNA pseudouridylate synthase domain containing 4
Gene Aliases
2410001E19Rik
Chromosome Location
Chr.9: 35267881 - 35275957 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028040.2 NP_082316.1 3 - 4 565 60
GenBank mRNA AK010322.1 - 3 - 4 582 60
AK156574.1 - 3 - 4 582 60
BC061256.1 - 3 - 4 566 60

View Full Product Details