The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146046.1 | NP_001139518.1 | 5 - 6 | 775 | 97 |
NM_001146047.1 | NP_001139519.1 | 6 - 7 | 774 | 97 | |
NM_145616.4 | NP_663591.3 | 6 - 7 | 973 | 97 | |
GenBank mRNA | AK049268.1 | - | 6 - 7 | 752 | 97 |
AK051325.1 | - | 5 - 6 | 580 | 97 | |
AK085022.1 | - | - | 768 | 97 | |
BC016574.1 | - | 5 - 6 | 542 | 97 |