Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pias1
Entrez Gene ID
56469
Gene Name
protein inhibitor of activated STAT 1
Gene Aliases
2900068C24Rik, Ddxbp1, GBP
Chromosome Location
Chr.9: 62878366 - 62980879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019663.3 NP_062637.2 11 - 12 1493 65
GenBank mRNA AF077950.1 - 11 - 12 1485 65
AK013753.1 - 7 - 8 827 65
AK075708.1 - 11 - 12 1494 65
AK153871.1 - 11 - 12 1498 65
BC051417.1 - 11 - 12 1490 65

View Full Product Details