Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ncdn
Entrez Gene ID
26562
Gene Name
neurochondrin
Gene Aliases
AU042419, MMS10-AE, Ms10ae, mKIAA0607, norbin
Chromosome Location
Chr.4: 126743750 - 126753566 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011986.4 NP_036116.3 1 - 2 458 78
GenBank mRNA AK154889.1 - 1 - 2 439 78
AK170502.1 - 2 - 3 173 78
BC017126.1 - 1 - 2 389 78
CD548169.1 - 1 - 2 430 78
CX244321.1 - 1 - 2 401 78

View Full Product Details