The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019715.2 | NP_062689.2 | 6 - 7 | 1241 | 74 |
GenBank mRNA | AK007070.1 | - | 6 - 7 | 1182 | 74 |
AK142436.1 | - | 6 - 7 | 998 | 74 | |
BC029325.1 | - | 6 - 7 | 1063 | 74 | |
BC043330.1 | - | 6 - 7 | 1022 | 74 |