Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdm5a
Entrez Gene ID
214899
Gene Name
lysine (K)-specific demethylase 5A
Gene Aliases
AA409370, C76986, Jarid1a, RBP2, Rbbp2
Chromosome Location
Chr.6: 120364099 - 120444574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145997.2 NP_666109.2 6 - 7 1299 72
GenBank mRNA AK159079.1 - 6 - 7 1276 72
AK166055.1 - 6 - 7 1280 72
BC010717.1 - 6 - 7 940 72
BC011506.3 - 6 - 7 1233 72
BC080691.1 - 6 - 7 985 72

View Full Product Details