The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199311.2 | NP_955015.1 | 1 - 2 | 232 | 92 |
GenBank mRNA | AY397675.1 | - | 1 - 2 | 167 | 92 |
BC049354.1 | - | 1 - 2 | 222 | 92 | |
CK626014.1 | - | 1 - 2 | 232 | 92 |