Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pef1
Entrez Gene ID
67898
Gene Name
penta-EF hand domain containing 1
Gene Aliases
2600002E23Rik, Peflin
Chromosome Location
Chr.4: 130107556 - 130128134 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026441.4 NP_080717.2 1 - 2 98 154
GenBank mRNA AK007394.1 - 1 - 2 40 154
AK013238.1 - 1 - 2 40 154
AK046247.1 - 1 - 2 40 154
AK075978.1 - 1 - 2 103 154
BC019191.1 - 1 - 2 46 154

View Full Product Details