Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pef1
Entrez Gene ID
67898
Gene Name
penta-EF hand domain containing 1
Gene Aliases
2600002E23Rik, Peflin
Chromosome Location
Chr.4: 130107556 - 130128134 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026441.4 NP_080717.2 2 - 3 372 59
GenBank mRNA AK007394.1 - 2 - 3 314 59
AK013238.1 - 2 - 3 314 59
AK046247.1 - 2 - 3 314 59
AK075978.1 - 2 - 3 377 59
BC019191.1 - 2 - 3 320 59

View Full Product Details