Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrtm4
Entrez Gene ID
243499
Gene Name
leucine rich repeat transmembrane neuronal 4
Gene Aliases
7530419J18Rik, A230052N11
Chromosome Location
Chr.6: 80018859 - 80810162 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134743.1 NP_001128215.1 1 - 2 305 58
NM_001282102.1 NP_001269031.1 1 - 2 305 58
GenBank mRNA AK038656.1 - 1 - 2 135 58
BC037216.1 - 1 - 2 227 58
BY130176.1 - 1 - 2 306 58


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CJ059229.1 - 1 - 2 216 Chr. 6 - 80018967 - 80021829 [+] on Build GRCm38 58
GenBankmRNA BI732750.1 - 1 - 2 228 Chr. 6 - 80018955 - 80022127 [+] on Build GRCm38 58
GenBankmRNA BB623691.1 - 1 - 2 192 Chr. 6 - 80018991 - 80022049 [+] on Build GRCm38 58
GenBankmRNA CJ058784.1 - 1 - 2 219 Chr. 6 - 80018964 - 80021794 [+] on Build GRCm38 58
GenBankmRNA BB631932.1 - 1 - 2 135 Chr. 6 - 80019048 - 80022074 [+] on Build GRCm38 58
GenBankmRNA CJ054519.1 - 1 - 2 288 Chr. 6 - 80018895 - 80021749 [+] on Build GRCm38 58
GenBankmRNA BY230277.1 - 1 - 2 228 Chr. 6 - 80018954 - 80021752 [+] on Build GRCm38 58
GenBankmRNA CJ059209.1 - 1 - 2 257 Chr. 6 - 80018926 - 80021789 [+] on Build GRCm38 58
GenBankmRNA CJ057204.1 - 1 - 2 218 Chr. 6 - 80018964 - 80021798 [+] on Build GRCm38 58
GenBankmRNA CJ048144.1 - 1 - 2 204 Chr. 6 - 80018978 - 80021842 [+] on Build GRCm38 58


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream predicted gene, 20594 Gm20594 Mtrnr2l -200.5kb Sense Chr. - 79818031 - 79818364 on Build GRCm38
Downstream microRNA 7232 Mir7232 mmu-mir-7232 1.9mb Anti-sense Chr. - 81895590 - 81895663 on Build GRCm38

View Full Product Details