Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il23r
Entrez Gene ID
209590
Gene Name
interleukin 23 receptor
Gene Aliases
IL-23R
Chromosome Location
Chr.6: 67422932 - 67491855 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144548.1 NP_653131.2 4 - 5 617 114
GenBank mRNA AF461423.1 - 4 - 5 617 114
BC112425.1 - 4 - 5 600 114

View Full Product Details