Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn1r45
Entrez Gene ID
22297
Gene Name
vomeronasal 1 receptor 45
Gene Aliases
V1RA9, V1r2, V1ra11, V1ra2, mV1R2
Chromosome Location
Chr.6: 89931650 - 89940507 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011684.2 NP_035814.2 1 - 2 148 157

View Full Product Details