Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Clec7a
- Entrez Gene ID
-
56644
- Gene Name
- C-type lectin domain family 7, member a
- Gene Aliases
-
BGR, Clecsf12, beta-GR
- Chromosome Location
-
Chr.6: 129461591 - 129472777 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AA240643.1 |
- |
2 - 3 |
273 |
72 |
View Full Product Details