The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034029.1 | NP_001029201.1 | 5 - 6 | 594 | 78 |
NM_001034031.1 | NP_001029203.1 | 7 - 8 | 669 | 78 | |
NM_145826.5 | NP_665825.2 | 6 - 7 | 1021 | 78 | |
GenBank mRNA | AF458068.1 | - | 6 - 7 | 671 | 78 |
AK046302.1 | - | 6 - 7 | 1024 | 78 | |
AK046713.1 | - | 6 - 7 | 683 | 78 | |
AK169498.1 | - | 6 - 7 | 849 | 78 | |
BC069861.1 | - | 6 - 7 | 698 | 78 | |
DQ092339.1 | - | 6 - 7 | 610 | 78 | |
DQ092340.1 | - | 5 - 6 | 594 | 78 | |
DQ092341.1 | - | 7 - 8 | 669 | 78 |