The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175313.4 | NP_780522.1 | 2 - 3 | 438 | 74 |
GenBank mRNA | AK037507.1 | - | 2 - 3 | 439 | 74 |