The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042556.1 | NP_001036021.1 | 9 - 10 | 1154 | 103 |
NM_023323.3 | NP_075812.3 | 9 - 10 | 804 | 103 | |
GenBank mRNA | AB041810.1 | - | 9 - 10 | 1184 | 103 |
AK010776.1 | - | 9 - 10 | 798 | 103 | |
AK011644.1 | - | 9 - 10 | 782 | 103 | |
AK012188.1 | - | 9 - 10 | 804 | 103 | |
AK012641.1 | - | 9 - 10 | 783 | 103 | |
AK013395.1 | - | 9 - 10 | 779 | 103 | |
AK017678.1 | - | 9 - 10 | 775 | 103 | |
AK020030.1 | - | 9 - 10 | 765 | 103 | |
AK051135.1 | - | 9 - 10 | 1155 | 103 |