Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map7
Entrez Gene ID
17761
Gene Name
microtubule-associated protein 7
Gene Aliases
E-MAP-115, MAP-7, Mtap7, R75000, mshi, mste, ste
Chromosome Location
Chr.10: 20148920 - 20281590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198635.1 NP_001185564.1 6 - 7 736 91
NM_008635.2 NP_032661.2 6 - 7 736 91
GenBank mRNA AB098611.1 - 6 - 7 642 91
AK008182.1 - 6 - 7 659 91
AK014575.1 - 6 - 7 653 91
AK133267.1 - 6 - 7 1185 91
BC042771.1 - 6 - 7 661 91
BC044733.1 - 6 - 7 661 91
BC046555.1 - 6 - 7 661 91
BC052637.1 - 6 - 7 672 91
Y15197.1 - 6 - 7 642 91

View Full Product Details