Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dmbx1
Entrez Gene ID
140477
Gene Name
diencephalon/mesencephalon homeobox 1
Gene Aliases
Atx, Cdmx, Mbx, Otx3
Chromosome Location
Chr.4: 115915119 - 115939926 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025567.1 NP_001020738.1 2 - 3 212 72
GenBank mRNA AB037698.1 - 2 - 3 193 72
AF421858.1 - 1 - 2 158 72
AF499446.1 - 1 - 2 1994 72
BC050912.1 - 2 - 3 189 72
BY732319.1 - 2 - 3 212 72

View Full Product Details