The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080924.2 | NP_001074393.1 | 7 - 8 | 1174 | 91 |
NM_001290501.1 | NP_001277430.1 | 8 - 9 | 1021 | 91 | |
GenBank mRNA | AK133342.1 | - | 8 - 9 | 1022 | 91 |
BC068259.1 | - | 2 - 3 | 265 | 91 | |
BC151080.1 | - | 8 - 9 | 895 | 91 | |
BC151083.1 | - | 8 - 9 | 895 | 91 |