The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025473.3 | NP_079749.1 | 1 - 2 | 469 | 77 |
GenBank mRNA | AK007715.1 | - | 1 - 2 | 470 | 77 |
BC011237.1 | - | 1 - 2 | 467 | 77 | |
BC019752.1 | - | 2 - 3 | 235 | 77 | |
BC054411.1 | - | 1 - 2 | 141 | 77 | |
BC069871.1 | - | 1 - 2 | 604 | 77 |