Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npc1l1
Entrez Gene ID
237636
Gene Name
NPC1-like 1
Gene Aliases
9130221N23Rik, Gm243
Chromosome Location
Chr.11: 6211011 - 6230245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207242.2 NP_997125.2 6 - 7 2282 91
GenBank mRNA AK078947.1 - 6 - 7 2183 91
AY437866.1 - 6 - 7 2160 91
BC131788.1 - 6 - 7 2159 91

View Full Product Details